Double open end semiconductor filter cartridge; 0.20 µm
Representative image only.
Item # WZ-06479-60
£141.00GBP / Each
£169.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolyethersulfone (PES)
  • Micron Rating0.2
  • Filter Length (in)10
  • Filtration Area (ft²)6.9
  • Filtration Area (in²)993.6
  • Filtration Area (m²)0.641
  • DescriptionDouble open end semiconductor filter cartridge; 0.20 µm
More About this Item
These cartridges are autoclavable to 250°F <121°C) and in-line sterilizable to 257°F (125°C) for up to 30-minute cycles. They are tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180°F (82°C) at 10 psid.
The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121°C Plastics guidelines, and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL.
Semiconductor Conductor Single-layer PES membrane over PP core. Each is flushed with high-purity water to leach less than 5 ppb TOC and less that 5 ppb trace metals. Ideal for any high-purity chemical or DI water application.
REVIEWS
Q&A
Does this filter produce ASTM Type 1 water quality?
This is just a filter, for polishing DI water. This is not a deionization cartridge or meant to replace such ionic exchange purification means. Please call for further assistance.
Jen
Is this the smallest sub-micron filter you carry? What housing would you suggest for this cartridge?
Matt
Please see part number 06479-48. It has a 0.1 micron rating. This filter is 10" in length so we would recommend any of the housings that fit a 10" filter. We offer a variety of filter housins so depending upon your compatibility needs, there are many options. Our general-purpose housing is part number 01508-25 (for a 10" filter). Please contact us to further discuss your application needs. (Contact information can be found under the Contact Us page on our website)
Jaime
What filter housings are compatible with this product?
Brent
There are many options available allowing you to choose based on housing material, flow rate, and connection size. Commonly 29820-00 (opaque blue) or 29820-10 (clear) housings are used. Both have 3/8" NPT(F) ports. Please contact us by phone if you are not sure which housing is correct for your application.
Zeke