This item is now discontinued. Please browse similar items
Digi-Sense Universal Industrial Thermocouple Probes Type J, 12" Length
Representative image only.
Digi-Sense Item # WZ-93830-74
A low cost alternative to thermowell assemblies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeJ
  • Min Temperature (° F)0
  • Max Temperature (° F)1400
  • Min Temperature (° C)-17
  • Max Temperature (° C)760
  • JunctionGrounded
  • Connector Type1/2\" NPT(M) x 3/4\" NPT(F) conduit
  • Sheath material446 Stainless Steel
  • Sheath Length (in)12
  • DescriptionUniversal Industrial Thermocouple Probes Type J, 12\" Length
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Uniwell eliminates the need for thermowells and spring loaded probes. With a unique adjustable feature that allows the probe to be customized to the required insertion length, through the use of an adjustable ½” NPT compression fitting. Also designed to be directly inserted into the medium being measured without the use of a thermowell. This allows for ease of installation where a thermowell is not required. The probes conductor is made of 14 gauge wire and is encased with Magnesium Oxide insulation, and has a 446 stainless steel heavy wall sheath producing an increased life span. Featuring a standard size aluminum protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT(F) opening on the side for easy connection to