This item is now discontinued. Please browse similar items
Digi-Sense Type K Ungrounded Industrial Thermocouple Probe, 12" L, 3/4" Connection
Representative image only.
Digi-Sense Item # WZ-93830-63
All-in-one, includes industrial probe head, element and thermowell
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)-32
  • Max Temperature (° F)1600
  • Min Temperature (° C)-35
  • Max Temperature (° C)871
  • JunctionUngrounded
  • Connector Type3/4\" NPT(M) x 3/4\" NPT(F) conduit
  • Handle Material316 stainless steel
  • Sheath Length (in)12
  • Sheath Diameter (in)0.75
  • DescriptionType K Ungrounded Industrial Thermocouple Probe, 12\" L, 3/4\" Connection
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Thermocouple Probes Assemblies are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces, and vessels. The probes conductor is made of 20 gauge wire encased by ceramic insulators along with a standard size aluminum protection head. Head features a gasketed cap to seal out dust and moisture and houses a ceramic terminal block with screw terminals. Head has a 3/4" NPT (F) opening on the side for easy connection to electrical conduit. 316 stainless steel thermowells are included in assemble and are ideal for applications where removing of the probe without draining the vessel is necessary.