This item is now discontinued. View the direct replacement: WZ-18527-07 or browse similar items
Digi-Sense Type K Thermocouple Std Conn; Connection, Jack; Color, Yellow; 10/Pk
Representative image only.
Digi-Sense Item # WZ-93840-63
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeStandard Connector
  • ColorYellow
  • Max Temperature (° F)400
  • Max Temperature (° C)204
  • DescriptionType K Thermocouple Std Conn; Connection, Jack; Color, Yellow; 10/Pk
  • Warranty1 month
1 Month Warranty
Key Features
  • 1 month warranty
More About this Item
Connectors withstand continuous temperature of 350degF (177degC) and maximum temperature of 400degF (204degC). Feature glass-filled nylon casing with finger grips and polarity indicators; negative terminals are marked with red discs for easy identifaction. Miniconnectors accept miniature cable clamps and cable up to 20 gauge; standard connectors accept standard cable clamps and cable up to 16 gauge.