This item is now discontinued. Please browse similar items
Digi-Sense Twin Temperature 3" Back Connect Face, 6" Stem, RTD Transmitter; 0-250F
Representative image only.
Digi-Sense Item # WZ-93820-49
Critical process data can be captured remotely while still allowing local monitoring by the operator
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeRTD
  • Min Temperature (° F)0
  • Max Temperature (° F)250
  • Min Temperature (° C)-20
  • Max Temperature (° C)120
  • Connector Type1/2\" NPT(M)
  • Sheath Length (in)6
  • DescriptionTwin Temperature 3\" Back Connect Face, 6\" Stem, RTD Transmitter; 0-250F
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Cole Parmer Twin Temp thermometers are the perfect solution for systems requiring local and remote temperature readings. Local readings are provided by either an analog gauge. The remote reading is achieved by a continuous 4-20 mA output signal. All sensors are located in the thermometer stem and will easily fit into existing bi-metal thermowells of corresponding length. Housings are manufactured out of rugged 304 stainless guaranteed hermetically sealed to prevent fogging.