This item is now discontinued. Please browse similar items
Traceable Universal Digital Stopwatch with Calibration
Representative image only.
Traceable Item # WZ-98766-08
Single unit with 8-point memory acts as a stopwatch, timer, pacer, and chronograph!
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Provides repeat function of countdown alarm timing
  • Set pacer function from 0.1 (600 beats/min) to 30 seconds (2 beats/min)—beat counting up to 99 times
  • Traceable® model number 1048
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity9 hours, 59 minutes, 59 seconds, 99 hundredths
  • Timing ModesTime-out/time-in, continuous timing, cumulative split, interval split, memories, sample counter, chronograph
  • Count DirectionUp and down
  • Number of Programs8-point memory
  • Resolution0.01 sec
  • Accuracy±0.001%
  • AlarmAudible
  • DisplayTwo-line LCD
  • CaseABS Plastic
  • CertificationsNIST-traceable
  • Battery Type3 V button cell
  • Battery IncludedYes
  • Height (in)2 3/8
  • Depth (in)1/2
  • Width (in)2 3/8
  • DescriptionTraceable® Universal Digital Stopwatch with Calibration
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Provides repeat function of countdown alarm timing
  • Set pacer function from 0.1 (600 beats/min) to 30 seconds (2 beats/min)—beat counting up to 99 times
  • Traceable® model number 1048
More About this Item
When zero is reached, the alarm sounds and repeats the countdown from the programmed time. Timing cycle repeats automatically, or until stopped; count area of display shows number of repeat cycles. The dual-line LCD continuously displays time remaining to zero—ideal for analysis that must be noted at routine intervals.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.