This item is now discontinued. We recommend the following as suggested replacements: WZ-08402-60
Digi-Sense Traceable® Remote Probe Digital Thermometer with Calibration; Stick-Style, Celsius
Representative image only.
Digi-Sense Item # WZ-90205-23
Remote sensor and cable can be submerged
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° C)-50
  • Max Temperature (° C)70
  • Resolution0.1°
  • Display Type0.4\" Display, 3.5-digit LCD
  • No. of Probe(s) Included1
  • Probe Length (in)1/5
  • Battery1.5v Battery (1)
  • DescriptionTraceable® Remote Probe Digital Thermometer with Calibration; Stick-Style, Celsius
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
Stick-Style Remote Probe Thermometers have a 3-ft flexible cord, Min/Max memory, and simultaneous display of probe and ambient temperatures. Meter is chemical-resistant and shockproof; comes with a remote sensor and cable that can be submerged. Use as a pocket thermometer or mount for continuous use with the supplied self-adhesive hook-and-loop tabs.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.