This item is now discontinued. We recommend the following as suggested replacements: WZ-94461-07
Digi-Sense Traceable® Digital Timer with Memory and Calibration
Representative image only.
Digi-Sense Item # WZ-94461-26
Simple three-button operation
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity24 hours
  • Count DirectionDown
  • Number of Channels1
  • Resolution1 min, 1 sec during the last 10 minutes of countdown
  • Alarm1 minute beep
  • DisplayLCD, 1/3\" H digits
  • Battery Type1.5 V AgO
  • Depth (in)1/2
  • Outside Diameter (OD) (in)2 3/8
  • Depth (cm)1.3
  • Outside Diameter (OD) (cm)6
  • DescriptionTraceable® Digital Timer with Memory and Calibration
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • 1 year warranty
More About this Item
Unique memory returns the display to the previously programmed time at the touch of a button. Loud, beeping alarm sounds for one minute. Pocket-sized unit features a clip, magnetic back, and stand for mounting flexibility.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.