Digi-Sense TI.ST Dual-Magnet Surface Thermometers; Range 0 - 500F
Representative image only.
Digi-Sense Item # WZ-08107-04
Inexpensive way to easily measure surface temperature
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
  • 1 year warranty
PRODUCT OPTIONS
£55.76GBP / Each
£66.91GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)0
  • Max Temperature (° F)500
  • Division (° F)5
  • Dial Size (in)2
  • Dial / Case Material304 Stainless Steel
  • Stem Material304 Stainless Steel
  • DescriptionDual-Magnet Surface Thermometers; Range 0 - 500F
  • Warranty1 year
1 Year Warranty
Key Features
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
  • 1 year warranty
More About this Item
An ideal choice for use on ovens, boilers, process lines, motors, or anywhere a surface temperature needs to be measured. The specially designed bimetal sensing element and housing provide quick readings. These thermometers feature a dual magnet mount that will attach to an ferrous surface, steel case, glass lens and polished aluminum 2" dial with easy-to-read graduations.