This item is now discontinued. Please browse similar items
Digi-Sense Thermowell, 304 Stainless Steel, 9" Length, 1/2" Connection
Representative image only.
Digi-Sense Item # WZ-90433-83
Designed for easy replacement of your industrial probe without shutting the operation down.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material304 Stainless Steel
  • Min Temperature (° F)-32
  • Max Temperature (° F)1600
  • Min Temperature (° C)0
  • Max Temperature (° C)871
  • DescriptionThermowell, 304 Stainless Steel, 9\" Length, 1/2\" Connection
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
To match your probe to a thermowell, first measure the distance from the probe’s tip to the top of the thread (typical thread length is 0.5" for 1/2" NPT fittings). Next, match this length to the probe depth of the thermowell (“A” dimension). Other thermowell sizes and materials are available—call our Application Specialists for more information.
Limited-Space Thermowell when you need a shorter insertion depth.
Standard Thermowell for general-purpose applications.
Heavy-Duty Thermowell for higher fluid velocity ratings.