Traceable Calibrated Replacement General Purpose Probe for Scientific Thermistor Thermometer
Representative image only.
Traceable Item # WZ-90080-18
Optimize performance of your thermistor thermometer
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Designed with USB connection
  • Probe features 6" stem with 0.4" diameter
  • Total probe length is 9" with handle and features a 66" long cable
  • Traceable® model number 4111
  • 1 year warranty
£88.00GBP / Each
£105.60GBP (incl VAT)
Time to recalibrate?
for an additional
£158.34GBP / Each
£190.01GBP (incl VAT)
for an additional
£217.08GBP / Each
£260.50GBP (incl VAT)
for an additional
£218.47GBP / Each
£262.16GBP (incl VAT)
for an additional
£233.74GBP / Each
£280.49GBP (incl VAT)
Traceable Calibrated Four-Channel Alarm Timer
£32.76GBP / Each
£39.31GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermistor
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)302
  • Min Temperature (° C)-50
  • Max Temperature (° C)150
  • OutputUSB
  • Probe Length (in)9
  • Cable Length (in)66
  • Stem Diameter (in)0.4
  • Stem Length (in)6
  • DescriptionCalibrated Replacement General Purpose Probe for Traceable Scientific Thermistor Thermometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Designed with USB connection
  • Probe features 6" stem with 0.4" diameter
  • Total probe length is 9" with handle and features a 66" long cable
  • Traceable® model number 4111
  • 1 year warranty
More About this Item
These Traceable® thermistor probes are designed to be used with thermistor thermometer 90080-09. Featuring a temperature range of –58 to 302°F (–50 to 150°C), these probes are available in general-purpose or micro diameter options.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
For improved performance, probes should be calibrated as a system with the meter.