Traceable Calibrated Temperature Bluetooth® Enabled Data Logger Compatible with TraceableGO™ App; Ambient Sensor

Representative image only.
Traceable Item # WZ-18004-12
Easily access collected data between destinations with Bluetooth® data loggers
  • Connect to your data via smartphone or tablet—any Bluetooth-enabled device—using a downloadable Bluetooth App
  • View and download data with free TraceableGO™ App - data can be exported from the TraceableGO™ app and loaded to TraceableLIVE® cloud service*
  • Data loggers store up to 90 alarm events
  • Readings and setup parameters easily viewable on large 2" by 1" LCD
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Monitor ambient temperature environment
  • 1 year warranty

PRODUCT OPTIONS

£63.82GBP / Each
£76.58GBP (incl VAT)
Time to recalibrate?
for an additional
£218.47GBP / Each
£262.16GBP (incl VAT)
for an additional
£233.74GBP / Each
£280.49GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Number of Channels1
  • Width (InchShort)3 1/2
  • Input TypeTemperature; integrated sensor
  • Max Temperature (° F)158
  • Height (in)2
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Resolution (° F)0.1°
  • Temperature Accuracy (° F)±0.7°F
  • Temperature Resolution (° C)0.1°
  • Temperature Accuracy (° C)±0.4°C
  • Sampling Rate1 minute to 12 hours
  • Memory Size64K (65,536) data points, 7.5 months on 5-minute interval
  • InterfaceBluetooth
  • Battery Type2 AAA (included)
  • Display TypeMultiline LCD
  • DescriptionCalibrated Temperature Bluetooth® Enabled Data Logger Compatible with TraceableGO™ App and TraceableLIVE® Cloud Service; Ambient Sensor
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration

Key Features

  • Connect to your data via smartphone or tablet—any Bluetooth-enabled device—using a downloadable Bluetooth App
  • View and download data with free TraceableGO™ App - data can be exported from the TraceableGO™ app and loaded to TraceableLIVE® cloud service*
  • Data loggers store up to 90 alarm events
  • Readings and setup parameters easily viewable on large 2" by 1" LCD
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Monitor ambient temperature environment
  • 1 year warranty

More About this Item

Track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals, and food items during transport to any location—Bluetooth® loggers monitor shipments while in transit. Compact, durable data loggers fit almost anywhere—even inside a small cooler.

Configure the data loggers from your device using the TraceableGO™ App to start; stop; change memory mode, unit preference, alarm setting, logging interval; and enable/disable alarm. When all the parameters are set, simply press START on the logger to begin monitoring. Once the shipment is delivered, double press START/STOP on the logger to enable Bluetooth® and export the data to your device. You will get an immediate response to see if your contents have been exposed to conditions outside the set parameters.

Measurements are updated every 5 seconds with user-defined reading intervals from 1 minute to 12 hours in 1-minute steps. Logging duration is 227 days (7.5 months) on a 5-minute logging interval. Data loggers store up to 90 alarm events. Data logger displays current reading, Min/Max, and run time or alarm time. 18004-13 only: Display toggles between temperature and humidity measurements every 5 seconds.

You have the option to subscribe to cloud-based TraceableLIVE® service to upload data to cloud—ideal for tracking data from multiple loggers or to comply with regulatory agency requirements. The low-cost TraceableLIVE® subscription provides unlimited trip data to be saved on the cloud as well as the ability to send a report to an email.

Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Related Article: How to Safely Transport Vaccines, how to meet the critical challenge of ensuring a temperature-controlled supply chain from manufacturer to end-user.

Guaranteed to have at least one year of calibration remaining on certificate.

*TraceableLIVE® subscription required to upload data from the TraceableGO™ app.

For sale only in Canada, Europe, and USA.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat