This item is now discontinued. Please browse similar items
Digi-Sense Sec. Ref. PRT, 0.187" x 9" , -60~300C, 5-Pin Din
Representative image only.
Digi-Sense Item # WZ-90452-39
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5615 and 5616
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeSecondary Reference PRT/RTD
  • Min Temperature (° F)-76
  • Max Temperature (° F)572
  • Min Temperature (° C)-60
  • Max Temperature (° C)300
  • Temperature Accuracy±0.015°C and ±0.01°C/100 hr
  • Response Time (Sec)9
  • Connector Type5-Pin DIN
  • Sheath Length (in)9
  • Sheath Diameter (in)0.1875
  • DescriptionSec. Ref. PRT, 0.187\" x 9\" , -60~300C, 5-Pin Din
  • Warranty1 year
1 Year Warranty
Key Features
  • Equivalent to Fluke Calibration / Hart Scientific Model 5615 and 5616
  • 1 year warranty
More About this Item
AM1730—Secondary reference PRT/RTD probes: These probes are the workhorse of laboratories. With a calibration uncertainty at 0°C of just ±0.015°C, these probes fully meet ITS-90 criteria for reference thermometers fully. The unique design achieves an excellent balance between hysteresis effect and mechanical/thermal shocks. Both series boast very low short term stability of 7 mK at TPW. The AM1730 has a range of -200 to 420°C.
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.