This item is now discontinued. We recommend the following as suggested replacements: WZ-90260-25
Digi-Sense Precision Liquid-In-Glass Thermometer; 30 to 394F, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # WZ-90260-32
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy included
Monitor your critical environments
Traceable® Jumbo-Display Refrigerator/Freezer Thermometers with Calibration
£79.57 - £92.37GBP / Each
£95.48 - £110.84GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)30
  • Length (\")24
  • Max Temperature (° F)394
  • Accuracy±2.0°F; ±3.0°F above 212°F
  • Division (° F)0.5
  • Thermometer FillBlue Spirit
  • Overall Length (in)24
  • Overall Length (mm)610
  • Immersion Depth (mm)610
  • Stem MaterialWhite-Back Glass
  • DescriptionPrecision Liquid-In-Glass Thermometer; 30 to 394F, Total Immersion, Organic Liquid Fill
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy included
More About this Item
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Fractional-degree thermometers offer higher precision. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.