Digi-Sense Precision Liquid-In-Glass Thermometer; -1 to 51C, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # WZ-90260-23
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
PRODUCT OPTIONS
£71.27GBP / Each
£85.52GBP (incl VAT)
£176.83 - £235.37GBP / Each
£212.20 - £282.44GBP (incl VAT)
for an additional
£176.83GBP / Each
£212.20GBP (incl VAT)
for an additional
£235.37GBP / Each
£282.44GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")17 29/32
  • Min Temperature (° C)-1
  • Max Temperature (° C)51
  • Accuracy±0.6°C
  • Division (° C)0.1
  • Thermometer FillBlue Spirit
  • Overall Length (in)17.9
  • Overall Length (mm)455
  • Immersion Depth (mm)455
  • Stem MaterialWhite-Back Glass
  • DescriptionPrecision Liquid-In-Glass Thermometer; -1 to 51C, Total Immersion, Organic Liquid Fill
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy
More About this Item
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Fractional-degree thermometers offer higher precision. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.