This item is now discontinued. Please browse similar items
Digi-Sense Prec. Ind. PRT, 0.25" x 12" , -200~670C, 5-Pin Din
Representative image only.
Digi-Sense Item # WZ-90452-31
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePrecision Industrial PRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1238
  • Min Temperature (° C)-200
  • Max Temperature (° C)670
  • Temperature Accuracy±0.05°C and ±0.04°C/100 hr
  • Response Time (Sec)5
  • Connector Type5-Pin DIN
  • Sheath Length (in)12
  • Sheath Diameter (in)0.25
  • DescriptionPrec. Ind. PRT, 0.25\" x 12\" , -200~670C, 5-Pin Din
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
AM1660 Series-High temperature precision industrial PRT/RTD probes: This is a high temperature version of the AM1640 probe. Manufactured using a coil suspension element design for increased shock and vibration resistancealong with flexibility and durability. The AM1660 has a temperature range of -200 to 670ºC (-328 to 1238ºF).
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.