This item is now discontinued. Please browse similar items
Digi-Sense Prec. Ind. PRT, 0.187" x 9" , -60~300C, Bare Wire
Representative image only.
Digi-Sense Item # WZ-90452-16
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePrecision Industrial PRT/RTD
  • Min Temperature (° F)-76
  • Max Temperature (° F)572
  • Min Temperature (° C)-60
  • Max Temperature (° C)300
  • Temperature Accuracy±0.05°C and ±0.04°C/100 hr
  • Response Time (Sec)5
  • Connector TypeStripped Wire
  • Sheath Length (in)9
  • Sheath Diameter (in)0.1875
  • DescriptionPrec. Ind. PRT, 0.187\" x 9\" , -60~300C, Bare Wire
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
AM1620 Series—Precision industrial PRT/RTD probes: Conform to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a coil suspension element design for increased shock and vibration resistancealong with flexibility and durability. All have an accuracy of 35 mK at 0°C and short-term stability of 10 mK at 0°C. The AM1620 has a range of -60 to 300ºC (-76 to 572ºF)
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.