This item is now discontinued. Please browse similar items
Cole-Parmer Irreversible 4-Point Micro Horizontal Temperature Label, 1241-270F; 10/Pk
Representative image only.
Cole-Parmer Item # WZ-08068-45
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators4
  • Max temperature (° F)270
  • Temperature Points (° F)241, 250, 261, 270
  • Initial colorWhite
  • First color changeBlack
  • Height (in)11/64
  • Width (in)1/2
  • DescriptionIrreversible 4-Point Micro Horizontal Temperature Label, 1241-270F; 10/Pk
  • Warranty2 year
Two year warranty
Key Features
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 2 year warranty
More About this Item
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.