Digi-Sense General Purpose Liquid-In-Glass Thermometer, 0 to 300°F, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # WZ-90260-28
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
PRODUCT OPTIONS
£21.03GBP / Each
£25.24GBP (incl VAT)
£176.83 - £235.37GBP / Each
£212.20 - £282.44GBP (incl VAT)
for an additional
£176.83GBP / Each
£212.20GBP (incl VAT)
for an additional
£235.37GBP / Each
£282.44GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments
Traceable® Jumbo-Display Refrigerator/Freezer Thermometers with Calibration
£79.57 - £92.37GBP / Each
£95.48 - £110.84GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12
  • Min Temperature (° F)0
  • Max Temperature (° F)300
  • Accuracy±3.0°F; ±4.0°F above 230°F
  • Division (° F)2
  • Thermometer FillBlue Spirit
  • Overall Length (in)12
  • Overall Length (mm)300
  • Immersion Depth (mm)300
  • Stem MaterialWhite-Back Glass
  • DescriptionGeneral Purpose Liquid-In-Glass Thermometer, 0 to 300°F, Total Immersion, Organic Liquid Fill
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy
More About this Item
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.