This item is now discontinued. Please browse similar items
Digi-Sense General Purpose Liquid-In-Glass Thermometer; 0 to 230F, 76mm Immersion, Organic Liquid Fill SKU 9026011
Representative image only.
Digi-Sense Item # WZ-90260-11
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
Monitor your critical environments
Traceable® Jumbo-Display Fridge/Freezer Digital Thermometers with Calibration
£67.11 - £74.83GBP / Each
£80.53 - £89.80GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12
  • Min Temperature (° F)0
  • Max Temperature (° F)230
  • Accuracy±3.0°F
  • Division (° F)2
  • Thermometer FillBlue Spirit
  • Overall Length (in)12
  • Overall Length (mm)300
  • Immersion Depth (mm)76
  • Stem MaterialWhite-Back Glass
  • DescriptionGeneral Purpose Liquid-In-Glass Thermometer; 0 to 230F, 76mm Immersion, Organic Liquid Fill SKU 9026011
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy
More About this Item
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.