This item is now discontinued. Please browse similar items
Digi-Sense Borescope, 960 x 240 Resolution
Representative image only.
Digi-Sense Item # WZ-20250-28
Ideal for inspecting duct work, wiring locations, piping, vehicles, and much more
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBorescope, 960 x 240 Resolution
  • Warranty2 year
Two year warrantyConformite EuropeenneIP 67
Key Features
  • 2 year warranty
More About this Item
Flexible camera-tipped probes provide easy viewing in hard-to-reach areas. Bright LCD monitor allows instant viewing of area being inspected. Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout.
Digi-Sense Borescope 100
* 3.0" color LCD for real-time viewing
* TV-Out hookup for real-time viewing on external video devices
* Four adjustable LED lights for operation in dark environments
* Waterproof (IP67-rated) flexible camera probe
* 180° image rotation on LCD
* Built-in Li battery with USB charging
* Languages: English or German