This item is now discontinued. Please browse similar items
Dickson TP425 USB Temperature/Humidity Datalogger
Representative image only.
Dickson Mfr # TP425 – Item # WZ-18010-01
Record up to 32,512 temperature and humidity data points
  • Push-to-start models give you control in data recording
  • Provides 21 CFR Part 11 compliance with optional software
  • Use software to calculate dew point and to create custom graphs
  • of your data
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature and humidity
  • Max Temperature (° F)158
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Resolution (° F)1
  • Temperature Accuracy (° F)±0.8 (over range +20 to +120); ±1.8 (< +20 and > +120)
  • Temperature Accuracy (° C)±0.5 (-6 to +48); ±1.0 (-6 to +48)
  • Display Type2.5 digit LCD
  • DescriptionUSB Temperature/Humidity Datalogger
  • Warranty1 year
1 Year Warranty
Key Features
  • Push-to-start models give you control in data recording
  • Provides 21 CFR Part 11 compliance with optional software
  • Use software to calculate dew point and to create custom graphs
  • of your data
  • 1 year warranty
More About this Item
Use these dataloggers to monitor food, chemical, drug, and equipment storage areas, clean rooms, incubators, and more! Units offer a push-to-start function that gives the operator more control of data recording. The easy to read LCD display allows you to view current and min/max readings with the push of a button and the red LED flashes to confirm sampling. The compact rugged design allows you to place these dataloggers just about anywhere.
The Windows® based software lets you set sampling intervals, view downloaded data, and create customized graphs and tables in seconds. It can even plot data from several loggers on a single graph to let you compare conditions. Software is required to program dataloggers; order separately below.