This item is now discontinued. Please browse similar items
Dickson SK500 Compact Temperature Data Loggers; 12/Pk
Representative image only.
Dickson Mfr # SK550 – Item # WZ-23033-40
Economical data loggers bundled in convenient 12 packs
  • Log conditions while goods are in storage or transit
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature
  • Max Temperature (° F)158
  • Min Temperature (° F)-4
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Accuracy (° F)±1.8
  • Temperature Accuracy (° C)±1.0
  • Memory Size3,840 data points
  • InterfaceRS-232
  • Battery Type1 x 3.6V lithium battery
  • Display TypeNone
  • Length (in)1 1/4
  • Height (in)1
  • Width (in)1/2
  • DescriptionCompact Temperature Data Loggers; 12/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Log conditions while goods are in storage or transit
  • 1 year warranty
More About this Item
This data logger is so compact that it can be used just about anywhere—place in shipping containers, refrigerators, freezers, coolers, ventilation ducts, or just about any place where temperature conditions need to be monitored. The delayed start feature allows how much time passes before logging begins. The customized graphical software calculates min/max and averages of all the data collected. The data
can also be exported to other programs such as Excel® or PowerPoint®. Software and cable sold separately