This item is now discontinued. Please browse similar items
Dickson Electronic Signal Data Logger
Representative image only.
Dickson Mfr # ES120 – Item # WZ-23030-30
Sets a new standard in accurate, easy-to-use, affordable PC based recording
  • Large storage capacity—up to 32,512 sample points!
  • Dual channels — measure and record current or voltage on either channel
  • Monitor readings in real time from your PC.
  • LED power status indicator
  • Automatically converts data into familiar engineering units
  • Use with any powered voltage or current transmitter with specified range.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeCurrent and voltage
  • Temperature Accuracy (° F)±1.8
  • Temperature Accuracy (° C)±1.0
  • Max Operating Temperature (° F)176
  • Min Operating Temperature (° F)-40
  • Battery Life5 years
  • Battery Type1 x 3V lithium battery (included); factory replaceable
  • Length (in)3 1/8
  • Height (in)2 1/8
  • Width (in)1/16
  • DescriptionElectronic Signal Data Logger
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Large storage capacity—up to 32,512 sample points!
  • Dual channels — measure and record current or voltage on either channel
  • Monitor readings in real time from your PC.
  • LED power status indicator
  • Automatically converts data into familiar engineering units
  • Use with any powered voltage or current transmitter with specified range.
  • 1 year warranty
More About this Item
The ES120 will allow simultaneous monitoring of multiple variables with one data logger. With little effort, pressure, air movement, flow, fluid level, electrical properties, moisture, gas concentrations and much more can be monitored. Due to its small size and rugged design, the ES120 can be placed just about anywhere. The ES120 is ideal for researchers, building operators, test & equipment engineers and quality control personnel. DicksonWare™ makes reading recorded data on your PC easy! Importing data into a Word document, Excel spreadsheet or PowerPoint presentation is a snap.