This item is now discontinued. Please browse similar items
Dibafit™ In-Line Connector, 1/4"-28 UNF(F) flat bottom, 1/16" OD tubing; 10/pk
Representative image only.
Diba Mfr # 002333 – Item # WZ-21940-05
Compact in size—ideal when space is limited
  • Constructed of durable PEEK
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")1/2
  • Fitting MaterialPEEK
  • Width (InchShort)25/64
  • Number Of Ports2
  • Connection TypeCompression x UNF(F)
  • Thread size UNF(F) (port 1) (in)1/4\"-28
  • ColorNatural
  • Orifice Dimensions (in)1/16
  • DescriptionDibafit™ In-Line Connector, 1/4\"-28 UNF(F) flat bottom, 1/16\" OD tubing; 10/pk
Key Features
  • Constructed of durable PEEK
More About this Item
Use these PEEK in-line couplings to quickly and easily connect tubing lines. First tubing line passes through the connector and is held securely in place by a gripper ferrule. The 1/4"-28 UNF(F) port accepts a standard threaded fitting from a second line.