Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 3.0 mm ID, PEEK™; 5/pk
Representative image only.
Diba Mfr # 008NB30-KD2L – Item # WZ-21941-38
Use adapters to quickly connect tubing lines or fittings
  • Easily connect soft tubing with a 1/4"-28 UNF(F) connection
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")27/32
  • Fitting MaterialPEEK
  • Width (InchShort)25/64
  • Number Of Ports2
  • Connection TypeBarb x UNF(F)
  • Thread size UNF(F) (port 2) (in)1/4\"-28
  • Barb Size / Tubing Id (Port 1) (mm)3
  • ColorNatural
  • DescriptionDibafit™ Adapter, 1/4\"-28 UNF(F) flat bottom to 3.0 mm ID, PEEK™; 5/pk
Key Features
  • Easily connect soft tubing with a 1/4"-28 UNF(F) connection
More About this Item
Designed for use with soft wall tubing, these barbed adapters have a superior barb with a crisp edge to hold the tubing securely in place. Select from acetal or PEEK material of construction.