This item is now discontinued. We recommend the following as suggested replacements: WZ-21941-34
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 2.5 mm ID, acetal; 5/pk
Representative image only.
Diba Mfr # 008NB25-AD2B – Item # WZ-21941-32
Use adapters to quickly connect tubing lines or fittings
  • Easily connect soft tubing with a 1/4"-28 UNF(F) connection
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal
  • Length (\")29/32
  • Width (InchShort)25/64
  • Number Of Ports2
  • Connection TypeBarb x UNF(F)
  • Thread size UNF(F) (port 2) (in)1/4\"-28
  • Barb Size / Tubing Id (Port 1) (mm)2.5
  • ColorNatural
  • DescriptionDibafit™ Adapter, 1/4\"-28 UNF(F) flat bottom to 2.5 mm ID, acetal; 5/pk
Key Features
  • Easily connect soft tubing with a 1/4"-28 UNF(F) connection
More About this Item
Designed for use with soft wall tubing, these barbed adapters have a superior barb with a crisp edge to hold the tubing securely in place. Select from acetal or PEEK material of construction.