This item is now discontinued. We recommend the following as suggested replacements: WZ-11942-67
Diba Omnifit® SolventPlus™ Fixed Column Endpiece, 10 mm; 1/ea
Representative image only.
Diba Mfr # 006SFE-10 – Item # WZ-11946-79
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)10
  • End CapFixed
More About this Item
SolventPlus columns accept both adjustable and fixed endpieces. Simply match the column ID to select the right size.