Diba Omnifit® O-Ring for Connector and Valve Caps, seal; 10/pk
Representative image only.
Diba Mfr # 001404 – Item # WZ-21939-46
Created tight seal, holds tube in place
  • Constructed of durable FKM
£101.21GBP / Pkg of 10
£121.45GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFKM
  • Compatible WithOmnifit® Cap Style Reducers 21939-07 and -17
  • DescriptionOmnifit® O-Ring for Connector and Valve Caps, seal; 10/pk
Key Features
  • Constructed of durable FKM
More About this Item
Use where chemical resistance is less important and when the exact OD of the tube is not known. O-rings compress into the cone angle in the body of the fitting so when the cap is tightened a seal is created and the tube is held in place.