This item is now discontinued. Please browse similar items
Diba Omnifit® HiT™ Fixed Column Endpiece, 10 mm; 1/ea
Representative image only.
Diba Mfr # 006HFE-10 – Item # WZ-11945-42
  • Mix and match endpieces to create a column for your application
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)10
  • End CapFixed
Key Features
  • Mix and match endpieces to create a column for your application
More About this Item
Each adjustable endpiece gives you the flexibility to modify the bed length up to 80 mm in height (up to 160 mm with two adjustable endpieces).