Diba Omnifit® EZ Column Replacement Glass, 10 mm ID x 330 mm L

Representative image only.
Diba Mfr # 006ERG-10-33 – Item # WZ-11942-36
Mix and match column length while using your existing endpieces for ultimate flexibility
  • Constructed of durable glass

PRODUCT OPTIONS

£190.25GBP / Each
£228.30GBP (incl VAT)
Include Column Today
Diba Omnifit® EZ Chromatography Columns with Two Adjustable Endpieces
£618.79 - £1,938.96GBP / Each
£742.55 - £2,326.75GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Column Length (mm)330
  • Column Inside Diameter (ID) (mm)10
  • Wetted MaterialsGlass
  • Compatable WithOmnifit® EZ and EZ SolventPlus™ Column 10 mm ID x 330 mm L
  • DescriptionOmnifit® EZ Column Replacement Glass, 10 mm ID x 330 mm L

Key Features

  • Constructed of durable glass

More About this Item

Use a different length tube with your existing endpieces to meet your specific requirements. Columns sold separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat