This item is now discontinued. Please browse similar items
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.056" ID x 0.122" OD, PEEK™; 25/pk
Representative image only.
Diba Mfr # 008MB142-310K2L – Item # WZ-21941-08
Both the ID and OD of tubing are firmly gripped for a secure connection
  • Low-carryover connection for flexible to semi-rigid tubing
  • Chemically inert PEEK™ construction
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Barb Size / Tubing Id (Port 1) (mm)1.42
  • Barb Size / Tubing Id (Port 1) (in)1/18
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.1
  • DescriptionMicroBarb® Adapter, 1/4\"-28 UNF(F) to 0.056\" ID x 0.122\" OD, PEEK™; 25/pk
Key Features
  • Low-carryover connection for flexible to semi-rigid tubing
  • Chemically inert PEEK™ construction
More About this Item
Use MicroBarb adapters to securely connect flexible and semi-rigid tubing. Adapters feature two components—barbed component to fit inside the tube ID and threaded outer connector that slips over the tubing OD to secure the barb in place—to create a female threaded flat-bottom port.