This item is now discontinued. Please browse similar items
Diba Chromatography Column Frit Kit, 6.6 mm dia
Representative image only.
Diba Mfr # 006FK-06 – Item # WZ-11944-83
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)6.6
More About this Item
Frit kit comes with two each of 10 µm PE, 10 µm PTFE, and 30 µm PTFE frits.