Diba Chromatography Column Frit, 35 mm dia, 20 µm, PTFE; 20/pk
Representative image only.
Diba Mfr # 006FR-35-20 – Item # WZ-11945-21
Select from a range of porosities for your media and chemistry
  • Extend the life of your columns with these replacement frits
PRODUCT OPTIONS
£162.56GBP / Pkg of 20
£195.07GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)35
  • Wetted MaterialsPTFE
  • Description20 µm
Key Features
  • Extend the life of your columns with these replacement frits
More About this Item
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit.