Diba Chromatography Column Frit, 25 mm dia, 50 µm, PTFE; 20/pk
Representative image only.
Diba Mfr # 006FR-25-50 – Item # WZ-11945-18
Select from a range of porosities for your media and chemistry
  • Extend the life of your columns with these replacement frits
PRODUCT OPTIONS
£105.98GBP / Pkg of 20
£127.18GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Column Inside Diameter (ID) (mm)25
  • Wetted MaterialsPTFE
  • Description50 µm
Key Features
  • Extend the life of your columns with these replacement frits
More About this Item
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit.