Diba BenchMark™ Microbore Fixed Column Endpiece, 3 mm, 50 µm PTFE frit; 4/pk
Representative image only.
Diba Mfr # 006BFE-03-50 – Item # WZ-11941-91
Frit comes pre-assembled with each endpiece
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 50 µm diameter/pore
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPTFE
  • Description50 µm
Key Features
  • Choose from PTFE or stainless steel construction
  • Fits microbore columns with 50 µm diameter/pore
More About this Item
The endpieces make the primary seal to glass microbore columns.
Ideally suited for Benchmark microbore glass columns.