This item is now discontinued. Please browse similar items
DeWalt DPG58-1D Reinforcer Safety Glasses, Black Frame, Clear lens
Representative image only.
DeWalt Mfr # DPG58-1D – Item # WZ-86497-26
Ergonomically positioned handgrip pattern on the temple for a secure grip
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1+ (High Impact)
  • DescriptionReinforcer Safety Glasses, Black Frame, Clear lens
More About this Item
Wraparound design offers side protection without distorting vision. Soft rubber nosepiece for added comfort. Rubber-tipped temples keep glasses in place. Polycarbonate lens is impact resistant and provides 99.9% UV protection.
Compliance: ANSI Z87.1+ (High Impact)