This item is now discontinued. Please browse similar items
Desco 17222 Toe Grounder with 1 MegaOhm Resistor
Representative image only.
Desco Mfr # 17222 – Item # WZ-26809-18
Great for tasks requiring mobility
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionToe Grounder with 1 MegaOhm Resistor
  • Warranty1 Year
1 Year WarrantyUL Listed.
Key Features
  • 1 year warranty
More About this Item
Heel and toe grounders help eliminate the possibility of ESD damage. Tear-resistant, dual-layer conductive rubber heel cups and toe bands have nonmarring interiors, and an RTG (resistance-to-ground) rating of 106 to 107 Ω. Hook and stretch loop fastening assemblies and straps provide an excellent fit for a wide range of shoe types.