This item is now discontinued. Please browse similar items
Desco 09070 Adjustable Elastic Wristband with 6' Cord
Representative image only.
Desco Mfr # 09070 – Item # WZ-26809-06
Protect delicate electronics from static damage
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdjustable Elastic Wristband with 6' Cord
  • Warranty1 Year
1 Year WarrantyUL Listed.
Key Features
  • 1 year warranty
More About this Item
Wristbands and straps have 4-mm stainless steel snap studs. Purchase with or without a coiled cord; cords are 6-ft long and include a 1-MΩ resistor. Cleanroom compatible and safe for microelectronics applications. One size fits all. UL-listed.
Speidel® Twist-O-Flex® Wristbands feature a 300-series stainless steel interior expansion band. Flexible design won't pinch or grab hair.
Elastic Adjustable Wristbands feature a wide, soft elastic band and 300-series stainless steel used for all metal parts.