Delmhorst F-6 F-6 Analog Hay Moisture Meter Only, Range 13% - 40%

Representative image only.
Delmhorst Mfr # F-6 – Item # WZ-59823-09
Prevent hay spoilage and fire hazards
  • Color-coded range at optimum moisture content shows at a glance if hay is safe to bale
  • Simplified controls are user friendly for entry-level use
  • Built-in calibration check ensures accurate readings
  • 1 year warranty

PRODUCT OPTIONS

£296.11GBP / Each
£355.33GBP (incl VAT)
Include Electrodes Today!
Delmhorst-Moisture Meter Electrodes
£64.00 - £382.02GBP / Each
£76.80 - £458.42GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Measurement TypePin
  • Moisture Range Non-Wood Scale (Min) (%)13
  • Moisture Range Non-Wood Scale (Max) (%)40
  • Display TypeAnalog
  • Battery1 9V (included)
  • Height (in)3
  • Width (in)8
  • Depth (in)1 5/8
  • DescriptionF-6 Analog Hay Moisture Meter Only, Range 13% - 40%
  • Warranty1 year
1 Year Warranty

Key Features

  • Color-coded range at optimum moisture content shows at a glance if hay is safe to bale
  • Simplified controls are user friendly for entry-level use
  • Built-in calibration check ensures accurate readings
  • 1 year warranty

More About this Item

This hay moisture meter is ideal for agricultural applications where you need to know if your crop is ready for baling and safe storage. Color-coding the optimum range of 18 to 22% MC (moisture content) lets you know at a glance if you need to let the hay dry further or if it needs to be wetted before baling.
Available in two ranges to suit requirements for both US domestic (13 to 40% MC) and export (6 to 30% MC) markets.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat