This item is now discontinued. Please browse similar items
CR Magnetics CR45-R CURRENT INDICATORRED LED
Representative image only.
CR Magnetics Mfr # CR45-R – Item # WZ-68480-64
Measures current flow quickly and inexpensively
  • Self Powered
  • Compact
  • Quick visual measurement
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • Self Powered
  • Compact
  • Quick visual measurement
  • 1 year warranty
More About this Item
These compact sensors are ideal for monitoring heater elements, observing remote loads, indicating phase loss, or monitoring motor operation conveniently and inexpensively. The conductor being monitored is routed though the current transformer window opening in the device which then provides power to the device and measures the current being monitored. The CRM1000 provides indication by illuminating a series of seven high efficiency LEDs relative to the current measured. The CRM2000 indicates current flow through a continuous light bar. The CRM3000 provides indication via an auto-ranging digital display that measures inputs up to 50A AC.
The CRM3000 can also sense remotely when used with the CR3110-1500-36 (sold separately below) and be configured to provide an alarm signal when used with the CRM3000-CBL-36 (sold separately below).