Corning Stripwell™ Plates, 96-Well, Polystyrene, High Binding; 100/Cs

Representative image only.
Corning Mfr # 2592 – Item # WZ-13020-00
Flat-bottom strips break apart and can be reassembled into strip holder
  • Strips have one-way fit into strip holder—prevents misorientation
  • Manufactured from polystyrene (PS)— ideal for uniform binding, high optical clarity, and low background absorption

PRODUCT OPTIONS

Call For Price
Include Lids Today!
Corning® 96-Well Sterile Microplate Lids, Polystyrene
£115.85 - £643.89GBP / Case of 100
£139.02 - £772.67GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)0.36
  • Well Bottom Diameter (mm)6.35
  • Well Top Diameter (mm)6.86
  • Well ShapeFlat
  • Lid (Included)No
  • SterileNo
  • Plate TypePlate
  • Plate ColorClear
  • Binding PropertyHigh
  • MaterialPolystyrene
  • Height (in)9/16
  • Width (in)3 3/8
  • Length (in)5 1/40
  • DescriptionPlates, 96-Well, Polystyrene, High Binding; 100/Cs

Key Features

  • Strips have one-way fit into strip holder—prevents misorientation
  • Manufactured from polystyrene (PS)— ideal for uniform binding, high optical clarity, and low background absorption

More About this Item

The Stripwell™ plates and strips are 1 x 8 well flat bottom strips designed to easily break apart to be placed into strip holder. Plates include strips and strip holder preassembled in a 96-well format. Strips are sold without a strip holder, holder 13020-50 sold separately. High and medium binding surface options available for a wide range of applications.

High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions— ideal for immunoassays. Binding capacity is approximately 400 to 500ng IgG/cm².

Medium binding surface is hydrophobic in nature and binds biomolecules through passive interactions and is suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface. Binding capacity is approximately 100 to 200ng IgG/cm².

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat