This item is now discontinued. Please browse similar items
Corning Standard Chromatography Column; 19 mm ID, 250 mmL, 24/40 Joint
Representative image only.
Corning Mfr # LG4568T-114 – Item # WZ-34695-36
Ideal for adsorption, gel, permeation, and ion exchange techniques
  • Constructed of durable borosilicate glass
SPECS & DESCRIPTION
Specifications & Description
  • SPE Cartridge Length (mm)1000
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)19
  • Fritted DiskNo
  • TS Joint 24/40Yes
  • DescriptionStandardChromatographyColumn;19mmID,250mmL,24/40joint
Key Features
  • Constructed of durable borosilicate glass
More About this Item
LabGlass Standard Chromatography Columns are made of borosilicate glass and are available with or without TS joints and fritted disks. The TS joints on top of columns permit the connection of a funnel or other reservoir. Fritted disks (40 to 60 µm porosity) support packing materials.