Labglass Standard Chromatography Column, Glass, No 24/40 Joint, No Fritted Disk, 10.5 mm ID x 250 mm L
Representative image only.
Labglass Mfr # LG4566T-104 – Item # WZ-34695-48
Ideal for adsorption, gel, permeation, and ion exchange techniques
  • Constructed of durable borosilicate glass
PRODUCT OPTIONS
£146.84GBP / Case of 1
£176.21GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)10.5
  • Wetted MaterialsGlass
  • Fritted DiskNo
  • TS Joint 24/40No
  • DescriptionStandard Chromatography Column, Glass, No 24/40 Joint, No Fritted Disk, 10.5 mm ID x 250 mm L
Key Features
  • Constructed of durable borosilicate glass
More About this Item
LabGlass Standard Chromatography Columns are made of borosilicate glass and are available with or without TS joints and fritted disks. The TS joints on top of columns permit the connection of a funnel or other reservoir. Fritted disks (40 to 60 µm porosity) support packing materials.