This item is now discontinued. Please browse similar items
Corning Gosselin Sterile Petri Dish w/ Frosted Area, double bagged, traceability labels; 825/pk
Representative image only.
Corning Gosselin Mfr # BP93D-102 – Item # WZ-34551-85
Culture dishes for any application
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCrystal polystyrene w/ frosted area
  • ClosureVented, automation-compatible
  • Height (mm)15
  • Diameter (mm)100
  • SterileYes
  • DescriptionSterile Petri Dish w/ Frosted Area, double bagged, traceability labels; 825/pk
Key Features
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
More About this Item
Traceable standard petri dishes are available in nonsterile and sterile (SAL 10⁻⁶) versions, crystal or optically clear polystyrene, and feature cleanroom packaging with two outer bags and bar code label.