This item is now discontinued. Please browse similar items
Corning Gosselin EPI1B-05 Sterile Straw Pipettes, 1 mL, 150 mm, 100/Bag; 12,000/Cs
Representative image only.
Corning Gosselin Mfr # EPI1B-05 – Item # WZ-34551-55
Easily reach deep into narrow bottles, vials, and blender bags
  • Ideal for primary dilution sample testing
  • Disposable polypropylene is sterilized to reduce cross-contamination
  • Wide diameter easily handles viscous liquids such as cream and oils
  • Packed in tamper-evident bags with product traceability
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • AutoclavableNo
  • Max Volume (mL)1
  • Overall Length (mm)150
  • Number Of Channels1
  • Wetted MatterialPolypropylene (PP)
  • Package Of100/Bag
  • DescriptionSterile Straw Pipettes, 1 mL, 150 mm, 100/Bag; 12,000/Cs
Sterile
Key Features
  • Ideal for primary dilution sample testing
  • Disposable polypropylene is sterilized to reduce cross-contamination
  • Wide diameter easily handles viscous liquids such as cream and oils
  • Packed in tamper-evident bags with product traceability
More About this Item
Disposable straw pipettes are ideal for transferring tiny amounts of liquids without the need for an exact volume.
Sterilized PP pipettes are packed individually in a tamper-evident bag, 25 pipettes per resealable bag, 40 bags per box, 12 boxes per case.