This item is now discontinued. Please browse similar items
Corning Gosselin Autoclavable Bags, 3L, polypropylene, with biohazard symbol; 500/cs
Representative image only.
Corning Gosselin Mfr # SA03-10 – Item # WZ-34551-93
Safely decontaminate waste in your autoclave
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")10
  • MaterialPP
  • Volume (Liters)3
  • Height (in)15 51/64
  • Thickness (mil)0.04
  • Description Autoclavable Bags, 3L, Polypropylene, with Biohazard symbol, 500/pk
Key Features
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
More About this Item
Gosselin autoclavable PP bags are 40 µm thick—strong enough to hold your lab waste during the autoclaving process. Bags withstand temperatures up to 273°F (134°C).