This item is now discontinued. Please browse similar items
Corning Gosselin Autoclavable Bags, 33L, polypropylene, with biohazard symbol; 335/cs
Representative image only.
Corning Gosselin Mfr # SA33-10 – Item # WZ-34551-97
Safely decontaminate waste in your autoclave
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPP
  • Length (\")19 45/64
  • Volume (Liters)33
  • Height (in)31 1/2
  • Thickness (mil)0.04
  • Description Autoclavable Bags, 33L, Polypropylene, with Biohazard symbol, 335/pk
Key Features
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
More About this Item
Gosselin autoclavable PP bags are 40 µm thick—strong enough to hold your lab waste during the autoclaving process. Bags withstand temperatures up to 273°F (134°C).