This item is now discontinued. Please browse similar items
Corning Gosselin Autoclavable Bags, 14L polypropylene; 500/cs
Representative image only.
Corning Gosselin Mfr # SA14-01 – Item # WZ-34551-34
Safely decontaminate waste in your autoclave
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are transparent with no printing
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")15 3/4
  • MaterialPP
  • Volume (Liters)14
  • Height (in)23 19/32
  • Thickness (mil)0.04
  • Description Autoclavable Bags, Polypropylene, 14L, 500/pk
Key Features
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are transparent with no printing
More About this Item
Gosselin autoclavable PP bags are 40 µm thick—strong enough to hold your lab waste during the autoclaving process. Bags withstand temperatures up to 273°F (134°C).