Corning 4519-155 Dual-angled sidearm fittings for 3L Culture Flasks

Representative image only.
Corning Mfr # 4519-155 – Item # WZ-01837-42
  • Fits 3 L angled sidearm spinner flask

PRODUCT OPTIONS

£319.40GBP / Each
£383.28GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionDual-angled sidearm fittings for 3L Culture Flasks

Key Features

  • Fits 3 L angled sidearm spinner flask

More About this Item

Keep your ProCulture spinner flask running smoothly with these parts and accessories.

The dual fittings are comprised of a PET insert with a fluorocarbon O-ring, and a polypropylene sealing cap. They include two 1/4" OD stainless steel tubes which extend to the bottom of the flask.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat